Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indol-3-ylacetic acid hydrazide 95%

Supplier: AMBEED, INC
Warning

A207139-1G A207139-25G A207139-5G
76691-344EA 13 USD InStock
76691-344 76691-346 76691-348
Indol-3-ylacetic acid hydrazide 95%
Indol-3-ylacetic acid hydrazide
Precaución: for Research Use Only


Formula: C₁₀H₁₁N₃O
MW: 189.22 g/mol
Melting Pt: 198
MDL Number: MFCD00005634
CAS Number: 5448-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty