Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Guanidinobenzoic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A211798-100G A211798-10G A211798-25G A211798-500G A211798-5G
76691-520EA 31 USD InStock
76691-520 76691-522 76691-524 76691-526 76691-528
4-Guanidinobenzoic acid hydrochloride 97%
4-Guanidinobenzoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀ClN₃O₂
MDL Number: MFCD00040587
CAS Number: 42823-46-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty