Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(Trifluoromethyl)picolinic acid 98%

Supplier: AMBEED, INC
Warning

A213885-100G A213885-10G A213885-1G A213885-250MG A213885-25G A213885-5G
76691-570EA 184 USD InStock
76691-570 76691-572 76691-574 76691-576 76691-578 76691-580
6-(Trifluoromethyl)picolinic acid 98%
6-(Trifluoromethyl)picolinic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
MDL Number: MFCD02180819
CAS Number: 131747-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty