Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methyl(pentyl)amino)propanoic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A220187-100G A220187-1G A220187-25G A220187-5G A220187-500G
76691-776EA 114 USD InStock
76691-776 76691-778 76691-780 76691-782 77626-112
3-(Methyl(pentyl)amino)propanoic acid hydrochloride 97%
3-(Methyl(pentyl)amino)propanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₉H₂₀ClNO₂
MDL Number: MFCD08275216
CAS Number: 625120-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty