Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenyl)-2-methylpropanoic acid 98%

Supplier: AMBEED, INC
Warning

A223381-100MG A223381-1G A223381-250MG
76691-856EA 8 USD InStock
76691-856 76691-858 76691-860
2-(4-Bromophenyl)-2-methylpropanoic acid 98%
2-(4-Bromophenyl)-2-methylpropanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD02684220
CAS Number: 32454-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty