Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A223618-100MG A223618-1G A223618-250MG A223618-5G
76691-864EA 12 USD InStock
76691-864 76691-866 76691-868 76691-870
Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride 98%
Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
MDL Number: MFCD00237767
CAS Number: 64665-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty