Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)glycine-tert-butyl ester 98%

Supplier: AMBEED, INC
Warning

A223921-100G A223921-10G A223921-1G A223921-25G A223921-5G
76691-876EA 59 USD InStock
76691-876 76691-878 76691-880 76691-882 76691-884
N-(tert-Butoxycarbonyl)glycine-tert-butyl ester 98%
N-(tert-Butoxycarbonyl)glycine-tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
MDL Number: MFCD00191866
CAS Number: 111652-20-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty