Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Axitinib 98%

Supplier: AMBEED, INC
Danger

A226761-100MG A226761-1G A226761-1MG A226761-250MG A226761-5G
76691-942EA 38 USD InStock
76691-942 76691-944 76691-946 76691-948 76691-950
Axitinib 98%
Axitinib
Precaución: for Research Use Only


Formula: C₂₂H₁₈N₄OS
MW: 386.5 g/mol
MDL Number: MFCD09837898
CAS Number: 319460-85-0
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty