Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutyl phenyl ketone 98%

Supplier: AMBEED, INC
Warning

A233850-1G A233850-25G A233850-5G A233850-100G
76692-108EA 19 USD InStock
76692-108 76692-110 77600-874 77626-238
Cyclobutyl phenyl ketone 98%
Cyclobutyl phenyl ketone
Precaución: for Research Use Only


Formula: C₁₁H₁₂O
MW: 160.2124 g/mol
MDL Number: MFCD00001326
CAS Number: 5407-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty