Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyltri-n-octylammonium chloride 97%

Supplier: AMBEED, INC
Warning

A235797-100G A235797-10G A235797-25G A235797-500G
76692-156EA 22 USD InStock
76692-156 76692-158 76692-160 76692-162
Methyltri-n-octylammonium chloride 97%
Methyltri-n-octylammonium chloride
Precaución: for Research Use Only


Formula: C₂₅H₅₄ClN
MW: 404.16 g/mol
MDL Number: MFCD00011862
CAS Number: 5137-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty