Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cilnidipine 98%

Supplier: AMBEED, INC
Warning

A236779-100MG A236779-10G A236779-10MG A236779-1G A236779-250MG A236779-25G A236779-50MG A236779-5G A236779-100G A236779-1MG
76692-182EA 10 USD InStock
76692-182 76692-184 76692-186 76692-188 76692-190 76692-192 76692-194 77626-260 77626-256 77626-258
Cilnidipine 98%
Cilnidipine
Precaución: for Research Use Only


Formula: C₂₇H₂₈N₂O₇
MDL Number: MFCD00865853
CAS Number: 132203-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty