Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

11H-Benzo[a]fluoren-11-one 98%

Supplier: AMBEED, INC
Warning

A237140-100MG A237140-1G A237140-250MG A237140-5G
76692-196EA 48 USD InStock
76692-196 76692-198 76692-200 76692-202
11H-Benzo[a]fluoren-11-one 98%
11H-Benzo[a]fluoren-11-one
Precaución: for Research Use Only


Formula: C₁₇H₁₀O
MDL Number: MFCD00215959
CAS Number: 479-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty