Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyrimidamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A238193-1G A238193-5G A238193-250MG
76692-212EA 33 USD InStock
76692-212 76692-214 77626-266
Butyrimidamide hydrochloride 95%
Butyrimidamide hydrochloride


Formula: C₄H₁₁ClN₂
MDL Number: MFCD00054341
CAS Number: 3020-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty