Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl oxalyl chloride 98%

Supplier: AMBEED, INC
Danger

A239629-25G A239629-100G
76692-258EA 19 USD InStock
76692-258 77626-272
Ethyl oxalyl chloride 98%
Ethyl oxalyl chloride
Precaución: for Research Use Only


Formula: C₄H₅ClO₃
MDL Number: MFCD00000706
CAS Number: 4755-77-5
UN: 2920
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty