Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ciprofibrate 98%

Supplier: AMBEED, INC
Warning

A240639-100G A240639-100MG A240639-1G A240639-25G A240639-500G A240639-5G
76692-272EA 232 USD InStock
76692-272 76692-274 76692-276 76692-278 76692-280 76692-282
Ciprofibrate 98%
Ciprofibrate
Precaución: for Research Use Only


Formula: C₁₃H₁₄Cl₂O₃
MDL Number: MFCD00467135
CAS Number: 52214-84-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty