Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Fluorophenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A242168-100G A242168-10G A242168-25G A242168-500G
76692-308EA 33 USD InStock
76692-308 76692-310 76692-312 76692-314
(4-Fluorophenyl)acetic acid 98%
(4-Fluorophenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄F)CH₂COOH
MW: 154.14 g/mol
Boiling Pt: 264
MDL Number: MFCD00004343
CAS Number: 405-50-5
Merck Index: 13,04204
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty