Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl carbamate 98%

Supplier: AMBEED, INC
Warning

A243204-100G A243204-10G A243204-25G
76692-330EA 61 USD InStock
76692-330 76692-332 76692-334
Butyl carbamate 98%
Butyl carbamate
Precaución: for Research Use Only


Formula: C₅H₁₁NO₂
MDL Number: MFCD00007967
CAS Number: 592-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty