Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7H-Benzo[c]carbazole 97%

Supplier: AMBEED, INC
Warning

A244878-100MG A244878-10G A244878-1G A244878-250MG A244878-25G A244878-5G
76692-348EA 5 USD InStock
76692-348 76692-350 76692-352 76692-354 76692-356 76692-358
7H-Benzo[c]carbazole 97%
7H-Benzo[c]carbazole
Precaución: for Research Use Only


Formula: C₁₆H₁₁N
MDL Number: MFCD00215940
CAS Number: 205-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty