Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-N-cyclopropylbenzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A249394-10G A249394-5G
76692-430EA 90 USD InStock
76692-430 76692-432
4-Bromo-N-cyclopropylbenzenesulfonamide 98%
4-Bromo-N-cyclopropylbenzenesulphonamide
Precaución: for Research Use Only


Formula: C₉H₁₀BrNO₂S
MDL Number: MFCD00463486
CAS Number: 331950-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty