Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Di-9-carbazolylbenzene 96%

Supplier: AMBEED, INC
Warning

A252183-100MG A252183-1G A252183-250MG A252183-5G
76692-502EA 6 USD InStock
76692-502 76692-504 76692-506 76692-508
1,3-Di-9-carbazolylbenzene 96%
1,3-Di-9-carbazolylbenzene
Precaución: for Research Use Only


Formula: C₃₀H₂₀N₂
MW: 408.50 g/mol
MDL Number: MFCD09836836
CAS Number: 550378-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty