Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2,3,4,5-tetrafluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A263299-100G A263299-25G A263299-5G
76692-726EA 134 USD InStock
76692-726 76692-728 76692-730
Ethyl-2,3,4,5-tetrafluorobenzoate 98%
Ethyl 2,3,4,5-tetrafluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₆F₄O₂
MDL Number: MFCD02094158
CAS Number: 122894-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty