Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Fluorophenylthio)acetic acid 97%

Supplier: AMBEED, INC
Warning

A264949-1G A264949-25G A264949-5G
76692-760EA 13 USD InStock
76692-760 76692-762 76692-764
(4-Fluorophenylthio)acetic acid 97%
(4-Fluorophenylthio)acetic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂S
MW: 186.21
MDL Number: MFCD00219825
CAS Number: 332-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty