Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl(4-bromophenyl)sulfane 98%

Supplier: AMBEED, INC
Danger

A272797-100MG A272797-1G A272797-250MG A272797-5G
76692-864EA 15 USD InStock
76692-864 76692-866 76692-868 77626-420
Benzyl(4-bromophenyl)sulfane 98%
Benzyl(4-bromophenyl)sulfane
Precaución: for Research Use Only


Formula: C₁₃H₁₁BrS
MDL Number: MFCD00028019
CAS Number: 53136-21-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty