Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Hydroxyphenyl)piperazine 97%

Supplier: AMBEED, INC
Danger

A278499-25G A278499-5G
76692-984EA 19 USD InStock
76692-984 76692-986
1-(4-Hydroxyphenyl)piperazine 97%
1-(4-Hydroxyphenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O
MDL Number: MFCD00066156
CAS Number: 56621-48-8
UN: 3259
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty