Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminobenzene-1,3-disulfonic acid 97% (contains ~10% H₂O)

Supplier: AMBEED, INC
Danger

A280841-250MG A280841-5G
76693-024EA 12 USD InStock
76693-024 76693-026
4-Aminobenzene-1,3-disulfonic acid 97% (contains ~10% H₂O)
4-Aminobenzene-1,3-disulfonic acid
Precaución: for Research Use Only


Formula: C₆H₇NO₆S₂
MDL Number: MFCD00035903
CAS Number: 137-51-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty