Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid 97%

Supplier: AMBEED, INC
Danger

A281339-10G A281339-1G A281339-25G A281339-500G A281339-5G
76693-032EA 57 USD InStock
76693-032 76693-034 76693-036 76693-038 76693-040
4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid 97%
4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₄
MW: 212.2423 g/mol
MDL Number: MFCD18459359
CAS Number: 18720-35-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty