Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Dimethyl cyclobutane-1,2-dicarboxylate 98%

Supplier: AMBEED, INC
Warning

A281795-100MG A281795-1G A281795-250MG
76693-048EA 62 USD InStock
76693-048 76693-050 76693-052
cis-Dimethyl cyclobutane-1,2-dicarboxylate 98%
cis-Dimethyl cyclobutane-1,2-dicarboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂O₄
MW: 172.17 g/mol
MDL Number: MFCD00029075
CAS Number: 2607-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty