Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorobis(cyclooctene)iridium(I) dimer 98%

Supplier: AMBEED, INC
Warning

A282357-100MG A282357-250MG
76693-054EA 96 USD InStock
76693-054 76693-056 76693-058
Chlorobis(cyclooctene)iridium(I) dimer 98%
Chlorobis(cyclooctene)iridium(I) dimer
Precaución: for Research Use Only


Formula: C₃₂H₅₆Cl₂Ir₂
MDL Number: MFCD00213465
CAS Number: 12246-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty