Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-ethoxybenzoate 98%

Supplier: AMBEED, INC
Warning

A284495-100G A284495-10G A284495-5G
76693-110EA 140 USD InStock
76693-110 76693-112 76693-114
Methyl-4-ethoxybenzoate 98%
Methyl-4-ethoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.2
Boiling Pt: 75
MDL Number: MFCD00017264
CAS Number: 23676-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty