Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Cyclohexanedicarboxylic acid 95%

Supplier: AMBEED, INC
Danger

A284776-10G A284776-1G A284776-250MG A284776-100MG A284776-5G
76693-116EA 241 USD InStock
76693-116 76693-118 76693-120 77626-460 77626-462
1,1-Cyclohexanedicarboxylic acid 95%
1,1-Cyclohexanedicarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00094668
CAS Number: 1127-08-8
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty