Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Anthracen-9-yl)-N-methylmethanamine 97%

Supplier: AMBEED, INC
Warning

A286423-100MG A286423-1G A286423-250MG A286423-5G
76693-144EA 15 USD InStock
76693-144 76693-146 76693-148 77626-472
1-(Anthracen-9-yl)-N-methylmethanamine 97%
1-(Anthracen-9-yl)-N-methylmethanamine
Precaución: for Research Use Only


Formula: C₁₆H₁₅N
MDL Number: MFCD00009933
CAS Number: 73356-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty