Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

10-((tert-Butoxycarbonyl)amino)decanoic acid 98%

Supplier: AMBEED, INC
Warning

A286902-100MG A286902-1G A286902-250MG A286902-25G
76693-154EA 7 USD InStock
76693-154 76693-156 76693-158 76693-160
10-((tert-Butoxycarbonyl)amino)decanoic acid 98%
10-((tert-Butoxycarbonyl)amino)decanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₂₉NO₄
MW: 287.39 g/mol
MDL Number: MFCD00270351
CAS Number: 173606-50-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty