Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-His-OMe 97%

Supplier: AMBEED, INC
Warning

A291675-1G A291675-250MG A291675-25G A291675-5G
76693-236EA 28 USD InStock
76693-236 76693-238 76693-240 76693-242
Boc-His-OMe 97%
Boc-His-OMe
Precaución: for Research Use Only


Formula: C₁₂H₁₉N₃O₄
MW: 269.301 g/mol
MDL Number: MFCD00167635
CAS Number: 2488-14-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty