Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-(Trifluoromethoxy)phenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A293824-100G A293824-1G A293824-250MG A293824-25G A293824-5G A293824-10G
76693-278EA 235 USD InStock
76693-278 76693-280 76693-282 76693-284 76693-286 77626-506
(3-(Trifluoromethoxy)phenyl)acetic acid 97%
(3-(Trifluoromethoxy)phenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄(OCF₃))CH₂COOH
MDL Number: MFCD00082480
CAS Number: 203302-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty