Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-(Trifluoromethoxy)phenyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A305292-100G A305292-1G A305292-25G A305292-5G
76693-474EA 267 USD InStock
76693-474 76693-476 76693-478 76693-480
(2-(Trifluoromethoxy)phenyl)acetic acid 98%
(2-(Trifluoromethoxy)phenyl)acetic acid
Precaución: for Research Use Only


Formula: F₃COC₆H₄CH₂COOH
MW: 220.15
MDL Number: MFCD00236325
CAS Number: 220239-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty