Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Octanamidoacetic acid 97%

Supplier: AMBEED, INC
Warning

A312863-100G A312863-25G A312863-500G
76693-610EA 57 USD InStock
76693-610 76693-612 76693-614
2-Octanamidoacetic acid 97%
2-Octanamidoacetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₃
MDL Number: MFCD00154701
CAS Number: 14246-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty