Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzoylmethane 98%

Supplier: AMBEED, INC
Warning

A313139-1000G A313139-100G A313139-25G A313139-500G
76693-622EA 87 USD InStock
76693-622 76693-624 76693-626 77626-562
Dibenzoylmethane 98%
Dibenzoylmethane
Precaución: for Research Use Only


Formula: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
MW: 224.26 g/mol
Boiling Pt: 357
MDL Number: MFCD00003085
CAS Number: 120-46-7
Merck Index: 13,03033
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty