Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzothiazol-2-ylmethanol 98%

Supplier: AMBEED, INC
Warning

A314593-1G A314593-250MG A314593-5G
76693-662EA 35 USD InStock
76693-662 76693-664 76693-666
Benzothiazol-2-ylmethanol 98%
Benzothiazol-2-ylmethanol
Precaución: for Research Use Only


Formula: C₈H₇NOS
MDL Number: MFCD00226293
CAS Number: 37859-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty