Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1-adamantanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A315242-1G A315242-250MG A315242-100G
76693-670EA 29.71 USD InStock
76693-670 76693-672 76693-674 77571-888
Methyl-1-adamantanecarboxylate 97%
Methyl-1-adamantanecarboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₈O₂
MDL Number: MFCD01838519
CAS Number: 711-01-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty