Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl benzoate 99%

Supplier: AMBEED, INC
Warning

A316591-1000G A316591-100G A316591-25G A316591-500G
76693-684EA 17 USD InStock
76693-684 76693-686 76693-688 76693-690
Butyl benzoate 99%
Butyl benzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MDL Number: MFCD00009439
CAS Number: 136-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty