Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrimidine-2,4,5,6(1H,3H)-tetraone tetrahydrate 95%

Supplier: AMBEED, INC
Warning

A318997-10G A318997-25G A318997-5G
76693-740EA 64 USD InStock
76693-740 76693-742 76693-744
Pyrimidine-2,4,5,6(1H,3H)-tetraone tetrahydrate 95%
Pyrimidine-2,4,5,6(1H,3H)-tetraone tetrahydrate
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O₈
MDL Number: MFCD00149400
CAS Number: 6010-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty