Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetamidosuccinic acid 98%

Supplier: AMBEED, INC
Warning

A319131-10G A319131-250MG
76693-746EA 371 USD InStock
76693-746 76693-748
2-Acetamidosuccinic acid 98%
2-Acetamidosuccinic acid
Precaución: for Research Use Only


Formula: C₆H₉NO₅
MW: 175.14 g/mol
MDL Number: MFCD00063183
CAS Number: 2545-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty