Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-nitroanisole 95%

Supplier: AMBEED, INC
Warning

A329270-100G A329270-10G A329270-1G A329270-25G A329270-500G A329270-5G
76693-864EA 183 USD InStock
76693-864 76693-866 76693-868 76693-870 76693-872 76693-874
3-Methyl-2-nitroanisole 95%
3-Methyl-2-nitroanisole
Precaución: for Research Use Only


Formula: C₈H₉NO₃
MDL Number: MFCD00007179
CAS Number: 5345-42-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty