Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5-dimethoxybenzoic acid 98%

Supplier: AMBEED, INC
Warning

A334236-100G A334236-10G A334236-1G A334236-25G A334236-5G
76693-930EA 207 USD InStock
76693-930 76693-932 76693-934 76693-936 76693-938
2-Bromo-4,5-dimethoxybenzoic acid 98%
2-Bromo-4,5-dimethoxybenzoic acid
Precaución: for Research Use Only


Formula: C₉H₉BrO₄
MDL Number: MFCD00017542
CAS Number: 6286-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty