Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A340437-100G A340437-1G A340437-250MG A340437-25G A340437-5G
76693-956EA 623 USD InStock
76693-956 76693-958 76693-960 76693-962 76693-964
4-(3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl)benzoic acid 98%
4-(3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl)benzoic acid
Precaución: for Research Use Only


Formula: C₂₁H₁₅N₃O₄
MDL Number: MFCD09951804
CAS Number: 201530-41-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty