Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitroquinoline 95%

Supplier: AMBEED, INC
Warning

A343861-100G A343861-10G A343861-25G A343861-5G
76694-014EA 60 USD InStock
76694-014 76694-016 76694-018 76694-020
5-Nitroquinoline 95%
5-Nitroquinoline
Precaución: for Research Use Only


Formula: C₉H₆N₂O₂
MDL Number: MFCD00006790
CAS Number: 607-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty