Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,3S)-rel-Cyclopent-4-ene-1,3-diyl diacetate 98%

Supplier: AMBEED, INC
Warning

A352213-100MG A352213-1G A352213-250MG A352213-5G
76694-104EA 12 USD InStock
76694-104 76694-106 76694-108 76694-110
(1R,3S)-rel-Cyclopent-4-ene-1,3-diyl diacetate 98%
(1R,3S)-rel-Cyclopent-4-ene-1,3-diyl diacetate
Precaución: for Research Use Only


Formula: C₉H₁₂O₄
MDL Number: MFCD03002563
CAS Number: 54664-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty