Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,7-Dibromo-9,9-dimethylfluorene 98%

Supplier: AMBEED, INC
Warning

A354392-100G A354392-10G A354392-25G A354392-5G
76694-122EA 79 USD InStock
76694-122 76694-124 76694-126 76694-128
2,7-Dibromo-9,9-dimethylfluorene 98%
2,7-Dibromo-9,9-dimethylfluorene
Precaución: for Research Use Only


Formula: C₁₅H₁₂Br₂
MDL Number: MFCD00099471
CAS Number: 28320-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty