Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trimethoxysilyl)propyl acetate 95%

Supplier: AMBEED, INC
Warning

A356702-100G A356702-25G A356702-5G
76694-150EA 114 USD InStock
76694-150 76694-152 76694-154
3-(Trimethoxysilyl)propyl acetate 95%
3-(Trimethoxysilyl)propyl acetate
Precaución: for Research Use Only


Formula: C₈H₁₈O₅Si
MDL Number: MFCD00039834
CAS Number: 59004-18-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty