Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acarbose 98%

Supplier: AMBEED, INC
Warning

A357603-1G A357603-1MG A357603-250MG A357603-50MG A357603-5G
76694-168EA 33 USD InStock
76694-168 76694-170 76694-172 76694-174 76694-176
Acarbose 98%
Acarbose
Precaución: for Research Use Only


Formula: C₂₅H₄₃NO₁₈
MDL Number: MFCD00869592
CAS Number: 56180-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty